2-Bromobenzothiazole, 99%
2-Bromobenzothiazole, 99%
2-Bromobenzothiazole, 99%
Thermo Scientific Chemicals

2-Bromobenzothiazole, 99%

CAS: 2516-40-7 | C7H4BrNS | 214.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63286.03
also known as H63286-03
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.90
Save 7.25 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.90
Save 7.25 (14%)
Each
Specifications
CAS2516-40-7
Chemical Name or Material2-Bromobenzothiazole
ColorYellow
Flash Point>110°C (230°F)
Formula Weight214.08
View more
2-Bromobenzothiazole, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzothiazole, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids and bases.
RUO – Research Use Only

General References:

  1. VJ Majo.; J Prabhakaran.; JJ Mann.; JSD Kumar. An efficient palladium catalyzed synthesis of 2-arylbenzothiazoles. Tetrahedron letters.200344 (47), 8535-8537.
  2. Y Li.; YL Wang.; JY Wang. A simple iodine-promoted synthesis of 2-substituted benzothiazoles by condensation of aldehydes with 2-aminothiophenol. Chemistry Letters.200635 (4), 460-461.