L-Tryptophanamide hydrochloride, 95%
L-Tryptophanamide hydrochloride, 95%
L-Tryptophanamide hydrochloride, 95%
L-Tryptophanamide hydrochloride, 95%
Thermo Scientific Chemicals

L-Tryptophanamide hydrochloride, 95%

CAS: 5022-65-1 | C11H14ClN3O | 239.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63405.06
also known as H63405-06
Price (USD)
114.65
Special offer
Online exclusive
Ends: 31-Dec-2025
135.00
Save 20.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
114.65
Special offer
Online exclusive
Ends: 31-Dec-2025
135.00
Save 20.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS5022-65-1
IUPAC Name(2S)-2-amino-3-(1H-indol-3-yl)propanamide hydrochloride
Molecular FormulaC11H14ClN3O
InChI KeyWOBDANBSEWOYKN-FVGYRXGTSA-N
SMILESCl.N[C@@H](CC1=CNC2=CC=CC=C12)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (HPLC)≥94.0%
Identification (FTIR)Conforms
Proton NMRConsistent with structure
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only