2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole, 97%
2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole, 97%
2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole, 97%
Thermo Scientific Chemicals

2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole, 97%

CAS: 94944-70-4 | C9H14N2O5 | 230.22 g/mol
Have Questions?
Catalog NumberQuantity
H63416.MD
also known as H63416-MD
250 mg
Catalog number H63416.MD
also known as H63416-MD
Price (USD)
2,138.65
Online Exclusive
2,376.00
Save 237.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
2,138.65
Online Exclusive
2,376.00
Save 237.35 (10%)
Each
Chemical Identifiers
CAS94944-70-4
IUPAC Name1-{5-[(1R,2S,3R)-1,2,3,4-tetrahydroxybutyl]-1H-imidazol-2-yl}ethan-1-one
Molecular FormulaC9H14N2O5
InChI KeyCQSIXFHVGKMLGQ-BWZBUEFSSA-N
SMILESCC(=O)C1=NC=C(N1)[C@@H](O)[C@H](O)[C@H](O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormPowder or crystalline powder
Assay from Suppliers CofA≥94.0% (NMR)
Proton NMRConforms to structure
2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole used for treatment of autoimmune disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole used for treatment of autoimmune disorders.

Solubility
Soluble in distilled water, DMSO (>5 mg/ml), clear (warmed), and PBS pH 7.2 (∼0.15 mg/ml).

Notes
RUO – Research Use Only

General References:

  1. S J Gobin, R F Legg, A J Paine. The role of lymphocyte production and migration in the lymphopenia caused by 2-acetyl-4-tetrahydroxybutyl imidazole. International Journal of Immunopharmacology. 1992, 14(4), 687-697.
  2. S J Gobin, R F Legg, A J Paine. An update on sphingosine-1-phosphate and other sphingolipid mediators. Nature Chemical Biology. 2010, 6(7), 489-497.