2-Thioxo-3-[3-(trifluoromethyl)phenyl]-4-thiazolidinone, 95%
2-Thioxo-3-[3-(trifluoromethyl)phenyl]-4-thiazolidinone, 95%
2-Thioxo-3-[3-(trifluoromethyl)phenyl]-4-thiazolidinone, 95%
Thermo Scientific Chemicals

2-Thioxo-3-[3-(trifluoromethyl)phenyl]-4-thiazolidinone, 95%

CAS: 315-08-2 | C10H6F3NOS2 | 277.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H63447.MD
also known as H63447-MD
Price (USD)
64.65
Online Exclusive
72.10
Save 7.45 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
64.65
Online Exclusive
72.10
Save 7.45 (10%)
Each
Chemical Identifiers
CAS315-08-2
IUPAC Name2-sulfanylidene-3-[3-(trifluoromethyl)phenyl]-1,3-thiazolidin-4-one
Molecular FormulaC10H6F3NOS2
InChI KeyMRDJXDJVVCNDND-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC(=C1)N1C(=O)CSC1=S
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only