2-(Fmoc-amino)benzoic acid, 98%
2-(Fmoc-amino)benzoic acid, 98%
2-(Fmoc-amino)benzoic acid, 98%
Thermo Scientific Chemicals

2-(Fmoc-amino)benzoic acid, 98%

CAS: 150256-42-1 | C22H17NO4 | 359.381 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H63459.14
also known as H63459-14
Price (USD)
233.65
Online Exclusive
259.00
Save 25.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
233.65
Online Exclusive
259.00
Save 25.35 (10%)
Each
Chemical Identifiers
CAS150256-42-1
IUPAC Name2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)benzoate
Molecular FormulaC22H16NO4
InChI KeyCNAVPEPPAVHHKN-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC=CC=C1NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field

Solubility
Soluble in water or 1% acetic acid.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only