N-Succinimidyl N-methylcarbamate, 97%
N-Succinimidyl N-methylcarbamate, 97%
N-Succinimidyl N-methylcarbamate, 97%
Thermo Scientific Chemicals

N-Succinimidyl N-methylcarbamate, 97%

CAS: 18342-66-0 | C6H8N2O4 | 172.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H63566.03
also known as H63566-03
Price (USD)
46.60
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
46.60
Each
Chemical Identifiers
CAS18342-66-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay from Supplier's CofA≥96.0%
FormPowder or crystals or crystalline powder
Infrared spectrumConforms
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive, store away from moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. J Martinez; J Oiry; J L Imbach; F Winternitz. Activated N-nitrosocarbamates for regioselective synthesis of N-nitrosoureas. Journal of Medicinal Chemistry. 1982, 25(2), 178-182.
  2. Pradyumna Kumar Mishra; Venkata Raghuram Gorantla; Nabila Akhtar; Priyanka Tamrakar; Subodh Kumar Jain; and Kewal Krishan Mauda. Analysis of cellular response to isocyanate using N-succinimidyl N-methylcarbamate exposure in cultured mammalian cells. Environmental and Molecular Mutagenesis. 2009, 50(4), 328-336.