It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Malose Jack Mphahlele; Hugues Kadem Paumo; Ahmed M. El-Nahas; and Morad M. El-Hendawy. Synthesis and Photophysical Property Studies of the 2,6,8-Triaryl-4-(phenylethynyl)quinazolines. Molecules. 2014, 19(1), 795-818.
- Malose Jack Mphahlele; Hugues Kamdem Paumo; Lydia Rhyman; and Ponnadurai Ramasami. Synthesis and Photophysical Properties of Polycarbo-Substituted Quinazolines Derived from the 2-Aryl-4-chloro-6-iodoquinazolines. Molecules. 2015,20(8), 14656-14683.