(S)-2-(Boc-amino)butyric acid, 95%
(S)-2-(Boc-amino)butyric acid, 95%
(S)-2-(Boc-amino)butyric acid, 95%
Thermo Scientific Chemicals

(S)-2-(Boc-amino)butyric acid, 95%

CAS: 34306-42-8 | C9H17NO4 | 203.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H63686.06
also known as H63686-06
Price (USD)
152.65
Online Exclusive
169.00
Save 16.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
152.65
Online Exclusive
169.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS34306-42-8
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Dieter Seebach; Albert K. Beck; Hans G. Bossler; Christian Gerber; Soo Y Ko; C. William Murtiashaw; Reto Naef; Shin-Ichiro Shoda; drian Thaler; Manfred Krieger; and Roland Wenger. Modification of Cyclosporin A (CS): Generation of an enolate at the sarcosine residue and reactions with electrophiles. Helvetica Chimica Acta. 1993, 76(4), 1564-1590.
  2. Zsolt Bozsó; Gergely Tóth; Richard F. Murphy; Sándor Lovas. Difficulties in coupling to conformationally constrained aromatic amino acids. Letters in Peptide Science. 2000, 7(3), 157-163.