L-Aspartic acid di-tert-butyl ester hydrochloride, 98%
L-Aspartic acid di-tert-butyl ester hydrochloride, 98%
L-Aspartic acid di-tert-butyl ester hydrochloride, 98%
L-Aspartic acid di-tert-butyl ester hydrochloride, 98%
Thermo Scientific Chemicals

L-Aspartic acid di-tert-butyl ester hydrochloride, 98%

CAS: 1791-13-5 | C12H24ClNO4 | 281.777 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H63716.06
also known as H63716-06
Price (USD)
89.30
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
89.30
Each
Add to cart
Chemical Identifiers
CAS1791-13-5
IUPAC Name1,4-di-tert-butyl 2-aminobutanedioate hydrochloride
Molecular FormulaC12H24ClNO4
InChI KeyGVLZIMQSYQDAHB-UHFFFAOYNA-N
SMILESCl.CC(C)(C)OC(=O)CC(N)C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
L-Aspartic acid di-tert-butyl ester hydrochloride may be used in neurological comparison studies with structurally similar compounds such as L-Glutamic acid di-tert-butyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Aspartic acid di-tert-butyl ester hydrochloride may be used in neurological comparison studies with structurally similar compounds such as L-Glutamic acid di-tert-butyl ester.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only
W J Freed; E H Ghoz; S Crump. Convulsant properties of L-glutamic acid di-tert butyl ester. Neurobehavioral toxicology and teratology. 1985, 7(3), 275-278.