Methyl 3-amino-5-methylthiophene-2-carboxylate, 97%
Methyl 3-amino-5-methylthiophene-2-carboxylate, 97%
Methyl 3-amino-5-methylthiophene-2-carboxylate, 97%
Thermo Scientific Chemicals

Methyl 3-amino-5-methylthiophene-2-carboxylate, 97%

CAS: 76575-71-8 | C7H9NO2S | 171.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63719.06
also known as H63719-06
Price (USD)
515.65
Online Exclusive
573.00
Save 57.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
515.65
Online Exclusive
573.00
Save 57.35 (10%)
Each
Specifications
CAS76575-71-8
Chemical Name or MaterialMethyl 3-amino-5-methylthiophene-2-carboxylate
Formula Weight171.22
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Methyl 3-amino-5-methylthiophene-2-carboxylate is employed as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-amino-5-methylthiophene-2-carboxylate is employed as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only