6-Bromoindole-2-carboxylic acid, 97%
6-Bromoindole-2-carboxylic acid, 97%
6-Bromoindole-2-carboxylic acid, 97%
Thermo Scientific Chemicals

6-Bromoindole-2-carboxylic acid, 97%

CAS: 16732-65-3 | C9H6BrNO2 | 240.056 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H63799.03
also known as H63799-03
Price (USD)
110.65
Online Exclusive
123.00
Save 12.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
110.65
Online Exclusive
123.00
Save 12.35 (10%)
Each
Specifications
CAS16732-65-3
Chemical Name or Material6-Bromoindole-2-carboxylic acid
Formula Weight240.05
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only