Thiophene-2-boronic acid pinacol ester, 98%
Thiophene-2-boronic acid pinacol ester, 98%
Thiophene-2-boronic acid pinacol ester, 98%
Thiophene-2-boronic acid pinacol ester, 98%
Thermo Scientific Chemicals

Thiophene-2-boronic acid pinacol ester, 98%

CAS: 193978-23-3 | C10H15BO2S | 210.098 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H63819.06
also known as H63819-06
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS193978-23-3
IUPAC Name4,4,5,5-tetramethyl-2-(thiophen-2-yl)-1,3,2-dioxaborolane
Molecular FormulaC10H15BO2S
InChI KeyFFZHICFAHSDFKZ-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97.5%
Appearance (Color)White to pale cream
FormPowder or crystals or crystalline powder
Melting Point64.0-71.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only