1-[3-(Dimethylamino)propyl]-3-ethylurea, 97%
1-[3-(Dimethylamino)propyl]-3-ethylurea, 97%
1-[3-(Dimethylamino)propyl]-3-ethylurea, 97%
Thermo Scientific Chemicals

1-[3-(Dimethylamino)propyl]-3-ethylurea, 97%

CAS: 32897-26-0 | C8H19N3O | 173.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H63845.06
also known as H63845-06
Price (USD)
576.65
Special offer
Online exclusive
Ends: 30-Jun-2026
678.00
Save 101.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
576.65
Special offer
Online exclusive
Ends: 30-Jun-2026
678.00
Save 101.35 (15%)
Each
Chemical Identifiers
CAS32897-26-0
IUPAC Name3-[3-(dimethylamino)propyl]-1-ethylurea
Molecular FormulaC8H19N3O
InChI KeyNGJUYARYEXGDNN-UHFFFAOYSA-N
SMILESCCNC(=O)NCCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97%
Proton NMRConforms to structure
Carbon-13 NMRConforms to structure
Appearance (Color)Colorless to pale yellow
FormLiquid
1-[3-(Dimethylamino)propyl]-3-ethylurea is used in the preparation of cabergoline, which is a dopamine receptor used to treat Parkinsons syndrom.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-[3-(Dimethylamino)propyl]-3-ethylurea is used in the preparation of cabergoline, which is a dopamine receptor used to treat Parkinsons syndrom.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Kang, M.; Peng, D.; Zhang, Y.; Yang, Y.; He, L.; Yan, F.; Sun, S.; Fang, S.; Wang, P.; Zhang, Z. An electrochemical sensor based on rhodamine B hydrazide-immobilized graphene oxide for highly sensitive and selective detection of Cu(II). New. J. Chem. 2015, 39 (4), 3137-3144.
  2. Liu, K.; Zhang, S. Highly Efficient Synthesis of 1,3-Dihydroxy-2-carboxycarbazole and Its Neuroprotective Effects. ACS Med. Chem. Lett. 2015, 6 (8), 894-897.