2-Bromopyridine-5-carboxaldehyde, 95%
2-Bromopyridine-5-carboxaldehyde, 95%
2-Bromopyridine-5-carboxaldehyde, 95%
2-Bromopyridine-5-carboxaldehyde, 95%
Thermo Scientific Chemicals

2-Bromopyridine-5-carboxaldehyde, 95%

CAS: 149806-06-4 | C6H4BrNO | 186.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H63867.06
also known as H63867-06
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
114.00
Save 17.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
114.00
Save 17.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS149806-06-4
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
A useful synthetic intermediate. 6-Bromonicotinaldehyde is used in wittig reaction. Hydrodehalogenation of 6-bromonicotinaldehyde to produce alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful synthetic intermediate. 6-Bromonicotinaldehyde is used in wittig reaction. Hydrodehalogenation of 6-bromonicotinaldehyde to produce alcohol.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. 2-7°C Refrigerate.
RUO – Research Use Only

General References:

  1. Xi-Bo Chen,; Qiu-Peng Hu,; Qian-Jia Yuan,; Wei Ding,; Jiangmeng Ren,; Bu-Bing Zeng. Simultaneous dehalogenation and hydrogenation reduction of halogen-heteroaromatic aldehydes. Tetrahedron Letters. 2012, 53(53), 3798-3801.
  2. authors. title . Journal name. year of publication , volume issue , pg no.