1-(Methylsulfonyl)piperazine, 97%
1-(Methylsulfonyl)piperazine, 97%
1-(Methylsulfonyl)piperazine, 97%
1-(Methylsulfonyl)piperazine, 97%
Thermo Scientific Chemicals

1-(Methylsulfonyl)piperazine, 97%

CAS: 55276-43-2 | C5H13N2O2S | 165.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H63870.06
also known as H63870-06
Price (USD)
41.40
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
41.40
Each
Add to cart
Specifications
CAS55276-43-2
Chemical Name or Material1-(Methylsulfonyl)piperazine
Formula Weight164.23
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
1-(Methylsulfonyl)piperazine is employed as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Methylsulfonyl)piperazine is employed as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. H.-K. Fun; C. S. Yeap; C. S. Chidan Kumar; H. S. Yathirajan; and B. Narayana. 4-(Methyl­sulfon­yl)piperazin-1-ium chloride. Acta Cryst. 2010 66361-o362.
  2. Istvan Hermecz; Agnes Horvath; Zoltan Meszaros; Christine De Vos; Ludovic Rodriguez. Nitrogen bridgehead compounds. 44. New antiallergic 4H-pyrido[1,2-a]pyrimidin-4-ones. 4. J. Med. Chem. 1984 27(10), 1253-1259.