3-Chloro-6-iodopyridazine, 95%
3-Chloro-6-iodopyridazine, 95%
3-Chloro-6-iodopyridazine, 95%
Thermo Scientific Chemicals

3-Chloro-6-iodopyridazine, 95%

CAS: 135034-10-5 | C4H2ClIN2 | 240.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63879.06
also known as H63879-06
Price (USD)
242.65
Online Exclusive
269.00
Save 26.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
242.65
Online Exclusive
269.00
Save 26.35 (10%)
Each
Specifications
CAS135034-10-5
Chemical Name or Material3-Chloro-6-iodopyridazine
Formula Weight240.43
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Reacant in Suzuki-Miyaura coupling reaction. Reactant in the synthesis of 6-Chloropyridazine-3-carbonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reacant in Suzuki-Miyaura coupling reaction. Reactant in the synthesis of 6-Chloropyridazine-3-carbonitrile.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, heat.
RUO – Research Use Only
Karène Urgin,; Christophe Aubé,; Muriel Pipelier,; Virginie Blot,; Christine Thobie-Gautier,; Stéphane Sengmany,; Jacques Lebreton. Pd-Catalyzed Chemoselective Cross-Coupling Reaction of Triaryl- or Triheteroarylbismuth Compounds with 3,6-Dihalopyridazines. European Journal of Organic Chemistry. 2013, 2013 (1), 117-124.