1-Indoleacetic acid, 95%
1-Indoleacetic acid, 95%
1-Indoleacetic acid, 95%
Thermo Scientific Chemicals

1-Indoleacetic acid, 95%

CAS: 24297-59-4 | C10H9NO2 | 175.187 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H63935.MD
also known as H63935-MD
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Specifications
Beilstein142924
CAS24297-59-4
ChEBICHEBI:72814
Chemical Name or Material1-Indoleacetic acid
Formula Weight175.18
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only