1H-Indazole-3-carboxylic acid, 98%
1H-Indazole-3-carboxylic acid, 98%
1H-Indazole-3-carboxylic acid, 98%
Thermo Scientific Chemicals

1H-Indazole-3-carboxylic acid, 98%

CAS: 4498-67-3 | C8H6N2O2 | 162.148 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H64001.22
also known as H64001-22
Price (USD)
328.65
Special offer
Online exclusive
Ends: 30-Jun-2026
387.00
Save 58.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
328.65
Special offer
Online exclusive
Ends: 30-Jun-2026
387.00
Save 58.35 (15%)
Each
Chemical Identifiers
CAS4498-67-3
IUPAC Name1H-indazole-3-carboxylic acid
Molecular FormulaC8H6N2O2
InChI KeyBHXVYTQDWMQVBI-UHFFFAOYSA-N
SMILESOC(=O)C1=NNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only