4-Bromo-2,3-difluorobenzaldehyde, 98%
4-Bromo-2,3-difluorobenzaldehyde, 98%
4-Bromo-2,3-difluorobenzaldehyde, 98%
4-Bromo-2,3-difluorobenzaldehyde, 98%
Thermo Scientific Chemicals

4-Bromo-2,3-difluorobenzaldehyde, 98%

CAS: 644985-24-0 | C7H3BrF2O | 221.001 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64044.06
also known as H64044-06
Price (USD)
263.65
Online Exclusive
293.00
Save 29.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
263.65
Online Exclusive
293.00
Save 29.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS644985-24-0
IUPAC Name4-bromo-2,3-difluorobenzaldehyde
Molecular FormulaC7H3BrF2O
InChI KeyIMOLPSNRLZLWQR-UHFFFAOYSA-N
SMILESFC1=C(Br)C=CC(C=O)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Off-white
Assay (HPLC)>97.5%
4-Bromo-2,3-difluorobenzaldehyde is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,3-difluorobenzaldehyde is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only