2-(Trifluoromethyl)pyridine-5-carboxaldehyde, 95%
2-(Trifluoromethyl)pyridine-5-carboxaldehyde, 95%
2-(Trifluoromethyl)pyridine-5-carboxaldehyde, 95%
Thermo Scientific Chemicals

2-(Trifluoromethyl)pyridine-5-carboxaldehyde, 95%

CAS: 386704-12-7 | C7H4F3NO | 175.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64054.06
also known as H64054-06
Price (USD)
272.65
Special offer
Online exclusive
Ends: 30-Jun-2026
320.00
Save 47.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
272.65
Special offer
Online exclusive
Ends: 30-Jun-2026
320.00
Save 47.35 (15%)
Each
Chemical Identifiers
CAS386704-12-7
IUPAC Name6-(trifluoromethyl)pyridine-3-carbaldehyde
Molecular FormulaC7H4F3NO
InChI KeyMRPAGRCGPAXOGS-UHFFFAOYSA-N
SMILESFC(F)(F)C1=NC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Purity≥94.0%
FormPowder or crystals or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only