9-Phenylcarbazole-3-boronic acid, 98%
9-Phenylcarbazole-3-boronic acid, 98%
9-Phenylcarbazole-3-boronic acid, 98%
Thermo Scientific Chemicals

9-Phenylcarbazole-3-boronic acid, 98%

CAS: 854952-58-2 | C18H14BNO2 | 287.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64059.06
also known as H64059-06
Price (USD)
247.65
Online Exclusive
275.00
Save 27.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
247.65
Online Exclusive
275.00
Save 27.35 (10%)
Each
Chemical Identifiers
CAS854952-58-2
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
9-Phenylcarbazole-3-boronic acid is used in the synthesis of alkyl-functionalized organic dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Phenylcarbazole-3-boronic acid is used in the synthesis of alkyl-functionalized organic dyes.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Masami Kuriyama; Norihisa Hamaguchi; Keisuke Sakata; and Osamu Onomura. One-Pot Synthesis of Heteroaryl and Diheteroaryl Ketones through Palladium-Catalyzed 1,2-Addition and Oxidation. European Journal of Organic Chemistry. 20132013(16), 3378-3385.
  2. Chang-Hwa Jun; Yong-Jin Pu; Masahiro Igarashi; Takayuki Chiba; Hisahiro Sasabe; Junji Kido. A Donor-Acceptor-type Host Material for Solution-processed Phosphorescent Organic Light-emitting Devices Showing High Efficiency. European Journal of Organic Chemistry. 201443(12), 1935-1936.