6-Chloropyridine-2-carboxaldehyde, 97%
6-Chloropyridine-2-carboxaldehyde, 97%
6-Chloropyridine-2-carboxaldehyde, 97%
6-Chloropyridine-2-carboxaldehyde, 97%
Thermo Scientific Chemicals

6-Chloropyridine-2-carboxaldehyde, 97%

CAS: 54087-03-5 | C6H4ClNO | 141.554 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64129.03
also known as H64129-03
Price (USD)
101.65
Special offer
Online exclusive
Ends: 30-Jun-2026
119.00
Save 17.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
101.65
Special offer
Online exclusive
Ends: 30-Jun-2026
119.00
Save 17.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS54087-03-5
IUPAC Name6-chloropyridine-2-carbaldehyde
Molecular FormulaC6H4ClNO
InChI KeyXTRLIKXVRGWTKW-UHFFFAOYSA-N
SMILESClC1=CC=CC(C=O)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only