9,9'-Spirobifluorene, 98%
9,9'-Spirobifluorene, 98%
9,9'-Spirobifluorene, 98%
Thermo Scientific Chemicals

9,9'-Spirobifluorene, 98%

CAS: 159-66-0 | C25H16 | 316.403 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64133.06
also known as H64133-06
Price (USD)
170.65
Special offer
Online exclusive
Ends: 30-Jun-2026
200.00
Save 29.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
170.65
Special offer
Online exclusive
Ends: 30-Jun-2026
200.00
Save 29.35 (15%)
Each
Chemical Identifiers
CAS159-66-0
IUPAC Name9,9'-spirobi[fluorene]
Molecular FormulaC25H16
InChI KeySNFCXVRWFNAHQX-UHFFFAOYSA-N
SMILESC1=CC=C2C(=C1)C1=CC=CC=C1C21C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
A blue light emitting material in organic light emitting diodes (OLEDs). Also used as a blue-emitting material in electroluminescent devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A blue light emitting material in organic light emitting diodes (OLEDs). Also used as a blue-emitting material in electroluminescent devices.

Solubility
Soluble in common organic solvents. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. S. Tang; M. R. liu; P. Lu; H. xia; M. li; Z. Q. xie; F. Z. shen; C. Gu; H. P. wang; B. yang; and Y. G. ma. A Molecular Glass for Deep-Blue Organic Light-Emitting Diodes Comprising a 9,9'-Spirobifluorene Core and Peripheral Carbazole Groups. Advanced Functional Materials. 200717(15), 2869-2877.
  2. Jean-Hugues Fournier; Thierry Maris; and James D. Wuest. Molecular Tectonics. Porous Hydrogen-Bonded Networks Built from Derivatives of 9,9‘-Spirobifluorene. J. Org. Chem. 200469(6),1762-1775.