2-Amino-5-(trifluoromethyl)benzoic acid, 95%
2-Amino-5-(trifluoromethyl)benzoic acid, 95%
2-Amino-5-(trifluoromethyl)benzoic acid, 95%
Thermo Scientific Chemicals

2-Amino-5-(trifluoromethyl)benzoic acid, 95%

CAS: 83265-53-6 | C8H6F3NO2 | 205.136 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H64141.03
also known as H64141-03
Price (USD)
421.65
Online Exclusive
468.00
Save 46.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
421.65
Online Exclusive
468.00
Save 46.35 (10%)
Each
Chemical Identifiers
CAS83265-53-6
IUPAC Name2-amino-5-(trifluoromethyl)benzoic acid
Molecular FormulaC8H6F3NO2
InChI KeyGLCQUPLYYXSPQB-UHFFFAOYSA-N
SMILESNC1=C(C=C(C=C1)C(F)(F)F)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only