3-Carboxybenzeneboronic acid pinacol ester, 97%
3-Carboxybenzeneboronic acid pinacol ester, 97%
3-Carboxybenzeneboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

3-Carboxybenzeneboronic acid pinacol ester, 97%

CAS: 269409-73-6 | C13H17BO4 | 248.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H64195.06
also known as H64195-06
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
61.00
Save 9.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
61.00
Save 9.35 (15%)
Each
Chemical Identifiers
CAS269409-73-6
IUPAC Name3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Molecular FormulaC13H17BO4
InChI KeyOPWAPCOSDAFWFB-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only