2-Fluoro-5-nitropyridine, 98%
2-Fluoro-5-nitropyridine, 98%
2-Fluoro-5-nitropyridine, 98%
Thermo Scientific Chemicals

2-Fluoro-5-nitropyridine, 98%

CAS: 456-24-6 | C5H3FN2O2 | 142.089 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H64210.06
also known as H64210-06
Price (USD)
56.70
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
56.70
Each
Chemical Identifiers
CAS456-24-6
IUPAC Name2-fluoro-5-nitropyridine
Molecular FormulaC5H3FN2O2
InChI KeyXOZAJNLUAODXSP-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(F)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

General References:

  1. W. Gruber. Pyridine derivatives: part v. on fluoro-nitropyridines.Can. J. Chem.,1953,31(11), 1020-1024.
  2. Oleg D. Mitkin, et al. Dithiane- and Trithiane-Based Photolabile Scaffolds for Molecular Recognition.Org. Lett.,2001,12(3), 1841-1844.