Pyrene-1-boronic acid, 95%
Pyrene-1-boronic acid, 95%
Pyrene-1-boronic acid, 95%
Thermo Scientific Chemicals

Pyrene-1-boronic acid, 95%

CAS: 164461-18-1 | C16H11BO2 | 246.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64211.03
also known as H64211-03
Price (USD)
49.60
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
49.60
Each
Chemical Identifiers
CAS164461-18-1
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field. Also used as a intermediate for organic light-emitting diode(OLED).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field. Also used as a intermediate for organic light-emitting diode(OLED).

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Mitchell B. Lerner; Nicholas Kybert; Ryan Mendoza; Romain Villechenon; Manuel A. Bonilla Lopez; and A. T. Charlie Johnson. Scalable, non-invasive glucose sensor based on boronic acid functionalized carbon nanotube transistors. Appl. Phys. Lett. 201372183113.
  2. Bertrand Reuillard; Alan Le Goff; Michael Holzingera; and Serge Cosnier. Non-covalent functionalization of carbon nanotubes with boronic acids for the wiring of glycosylated redox enzymes in oxygen-reducing biocathodes. J. Mater. Chem. B. 20142(16), 2228-2232.