2,4-Dichloro-6-methylpyridine, 97+%
2,4-Dichloro-6-methylpyridine, 97+%
2,4-Dichloro-6-methylpyridine, 97+%
2,4-Dichloro-6-methylpyridine, 97+%
Thermo Scientific Chemicals

2,4-Dichloro-6-methylpyridine, 97+%

CAS: 42779-56-6 | C6H5Cl2N | 162.013 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64248.03
also known as H64248-03
Price (USD)
68.65
Special offer
Online exclusive
Ends: 30-Jun-2026
80.30
Save 11.65 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
68.65
Special offer
Online exclusive
Ends: 30-Jun-2026
80.30
Save 11.65 (15%)
Each
Add to cart
Chemical Identifiers
CAS42779-56-6
IUPAC Name2,4-dichloro-6-methylpyridine
Molecular FormulaC6H5Cl2N
InChI KeyWUGTXQVNSRFDNV-UHFFFAOYSA-N
SMILESCC1=NC(Cl)=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.0%
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Stanislav Rádl; Wieland Hafner; Petr Hezký; Ivan Krejčí; Jan Proška; and Jan Taimr. Molecular Modification of Anpirtoline, a Non-Opioid Centrally Acting Analgesic. Collect. Czech. Chem. Commun. 1999 64(2), 363-376.