5-Hydroxypyridine-2-carboxylic acid, 98+%
5-Hydroxypyridine-2-carboxylic acid, 98+%
5-Hydroxypyridine-2-carboxylic acid, 98+%
Thermo Scientific Chemicals

5-Hydroxypyridine-2-carboxylic acid, 98+%

CAS: 15069-92-8 | C6H5NO3 | 139.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H64265.03
also known as H64265-03
Price (USD)
34.70
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
34.70
Each
Chemical Identifiers
CAS15069-92-8
IUPAC Name5-hydroxypyridine-2-carboxylate
Molecular FormulaC6H4NO3
InChI KeyCGRRUFNHHQCLDZ-UHFFFAOYSA-M
SMILESOC1=CC=C(N=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>98.0%
Useful synthetic intermediate. Also intermediate for pharmaceutical application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful synthetic intermediate. Also intermediate for pharmaceutical application.

Solubility
Soluble in acetone, dimethyl sulfoxide , ethanol and methanol.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Michel Ouellet; Benoit Barbeau; and Michel J. Tremblay. p56lck, ZAP-70, SLP-76, and Calcium-regulated Effectors Are Involved in NF-κB Activation by Bisperoxovanadium Phosphotyrosyl Phosphatase Inhibitors in Human T Cells. Journal of Biological Chemistry. 1999 27435029-35036.
  2. MP Hegarty. The isolation and identifiction of 5-Hydroxypiperidine-2-carboxylic Acid from Leucaena glauca Benth. Australian Journal of Chemistry. 1957 10(4), 484 - 488.