3-Cyano-4-methoxy-2-pyridone, 95%, Thermo Scientific Chemicals
3-Cyano-4-methoxy-2-pyridone, 95%, Thermo Scientific Chemicals
3-Cyano-4-methoxy-2-pyridone, 95%, Thermo Scientific Chemicals
3-Cyano-4-methoxy-2-pyridone, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Cyano-4-methoxy-2-pyridone, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H64279.14
also known as H64279-14
25 g
Catalog number H64279.14
also known as H64279-14
Price (USD)
299.65
Online Exclusive
333.00
Save 33.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
299.65
Online Exclusive
333.00
Save 33.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS21642-98-8
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
Used as a intermediate for gimeracil and pharmaceutical application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a intermediate for gimeracil and pharmaceutical application.

Solubility
Soluble in Dimethyl sulfoxide.

Notes
Hydroscopic. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from water/moisture and oxidizing agent.
RUO – Research Use Only

General References:

  1. Ladislav Skursky; David Burleson; and George R. Waller. Interconversion of Ricinine and N-Demethylricinine in Senescent and Green Castor Plant Leaves. The Journal of Biological Chemistry. 19692443238-3242.
  2. G. R. Waller; and L. Skursky. Translocation and Metabolism of Ricinine in the Castor Bean Plant, Ricinus communis L. Plant Physiol. 197250(5), 622-626.