4-Fluoro-2-methylpyridine, 96%
4-Fluoro-2-methylpyridine, 96%
4-Fluoro-2-methylpyridine, 96%
Thermo Scientific Chemicals

4-Fluoro-2-methylpyridine, 96%

CAS: 766-16-5 | C6H6FN | 111.119 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64284.03
also known as H64284-03
Price (USD)
79.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
79.00
Each
Chemical Identifiers
CAS766-16-5
IUPAC Name4-fluoro-2-methylpyridine
Molecular FormulaC6H6FN
InChI KeyLEJYGRVERQVBLB-UHFFFAOYSA-N
SMILESCC1=NC=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>95.0%
It is used in the correlations with and limitations of the α scale of solvent hydrogen bond donor acidities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the correlations with and limitations of the α scale of solvent hydrogen bond donor acidities.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from air, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store
RUO – Research Use Only

General References:

  1. C. S. Giam, et al. Medium effects on the fluorine-19 magnetic resonance spectra of fluoropyridines.J. Am. Chem. Soc.,1973,95(10), 3235-3239.
  2. Robert W. Taft and Mortimer J. Kamlet. Linear solvation energy relationships. Part 4. Correlations with and limitations of the α scale of solvent hydrogen bond donor acidities.J. Chem. Soc., Perkin Trans.2.,1979,(12), 1723-1729.