Ethyl 2-amidinoacetate hydrochloride, 95%
Ethyl 2-amidinoacetate hydrochloride, 95%
Ethyl 2-amidinoacetate hydrochloride, 95%
Thermo Scientific Chemicals

Ethyl 2-amidinoacetate hydrochloride, 95%

CAS: 57508-48-2 | C5H11ClN2O2 | 166.605 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H64288.06
also known as H64288-06
Price (USD)
93.65
Online exclusive
104.00
Save 10.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
93.65
Online exclusive
104.00
Save 10.35
Each
Chemical Identifiers
CAS57508-48-2
IUPAC Name(1-amino-3-ethoxy-3-oxopropylidene)azanium
Molecular FormulaC5H11N2O2
InChI KeyHSDKTLKBDJXJQU-UHFFFAOYSA-O
SMILESCCOC(=O)CC(N)=[NH2+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Xueyu Qi, et al. Synthesis of Multisubstituted 2-Aminopyrroles/pyridines via Chemoselective Michael Addition/Intramolecular Cyclization Reaction.Org. Lett.,2014,16(16), 4186-4189.
  2. Yi-Fei Han, et al. An AAA-DDD Triply Hydrogen-Bonded Complex Easily Accessible for Supramolecular Polymers.Chem. Eur. J.,2014,20(51), 16980-16986.