2,4-Dichloro-6-methylphenol, 98%
2,4-Dichloro-6-methylphenol, 98%
2,4-Dichloro-6-methylphenol, 98%
Thermo Scientific Chemicals

2,4-Dichloro-6-methylphenol, 98%

CAS: 1570-65-6 | C7H6Cl2O | 177.024 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64312.03
also known as H64312-03
Price (USD)
48.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.10
Save 8.45 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
48.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.10
Save 8.45 (15%)
Each
Chemical Identifiers
CAS1570-65-6
IUPAC Name2,4-dichloro-6-methylphenol
Molecular FormulaC7H6Cl2O
InChI KeyWJQZZLQMLJPKQH-UHFFFAOYSA-N
SMILESCC1=CC(Cl)=CC(Cl)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Assay from Suppliers CofA≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only