1-Bromo-4-fluorobutane, 97%
1-Bromo-4-fluorobutane, 97%
1-Bromo-4-fluorobutane, 97%
Thermo Scientific Chemicals

1-Bromo-4-fluorobutane, 97%

CAS: 462-72-6 | C4H8BrF | 155.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H64317.06
also known as H64317-06
Price (USD)
136.65
Online Exclusive
152.00
Save 15.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
136.65
Online Exclusive
152.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS462-72-6
IUPAC Name1-bromo-4-fluorobutane
Molecular FormulaC4H8BrF
InChI KeyWMCUHRDQSHQNRW-UHFFFAOYSA-N
SMILESFCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
1-Bromo-4-fluorobutane may be employed for the synthesis of fluoroalkyl[2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]alkylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-fluorobutane may be employed for the synthesis of fluoroalkyl[2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]alkylamines.

Solubility
Sparingly soluble in water at 25°C 0.48 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. L Martarello, et. al. Synthesis and characterization of fluorinated and iodinated pyrrolopyrimidines as PET/SPECT ligands for the CRF1 receptor.Nucl Med Biol.,2001,28(2), 187-95.
  2. Mady MF and Kelland MA. Fluorinated Quaternary Ammonium Bromides: Studies on Their Tetrahydrofuran Hydrate Crystal Growth Inhibition and as Synergists with Polyvinylcaprolactam Kinetic Gas Hydrate Inhibitor.Energy and Fuels.,2013,27(9), 5175-81.