2-Bromo-5-chloropyridine, 98%
2-Bromo-5-chloropyridine, 98%
2-Bromo-5-chloropyridine, 98%
Thermo Scientific Chemicals

2-Bromo-5-chloropyridine, 98%

CAS: 40473-01-6 | C5H3BrClN | 192.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H64367.22
also known as H64367-22
Price (USD)
194.65
Online Exclusive
216.00
Save 21.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
194.65
Online Exclusive
216.00
Save 21.35 (10%)
Each
Chemical Identifiers
CAS40473-01-6
IUPAC Name2-bromo-5-chloropyridine
Molecular FormulaC5H3BrClN
InChI KeyBZUUVQCSPHPUQA-UHFFFAOYSA-N
SMILESClC1=CC=C(Br)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is also used in the synthesis of novel halopyridinylboronic acids and esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is also used in the synthesis of novel halopyridinylboronic acids and esters.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Francis H. Case, et al. The Synthesis of Certain Substituted 2,2'-Bipyridyls.J. Am. Chem. Soc.,1946,68(12), 2574-2577.
  2. Alexandre Bouillon, et al. Synthesis of novel halopyridinylboronic acids and esters. Part 4: Halopyridin-2-yl-boronic acids and esters are stable, crystalline partners for classical Suzuki cross-coupling.Tetrahedron.,2003,59(50), 10043-10049.