(1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane, 98%
(1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane, 98%
(1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane, 98%
Thermo Scientific Chemicals

(1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane, 98%

CAS: 87583-89-9 | C8H18N2 | 142.246 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H64396.14
also known as H64396-14
Price (USD)
521.65
Special offer
Online exclusive
Ends: 30-Jun-2026
613.00
Save 91.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
521.65
Special offer
Online exclusive
Ends: 30-Jun-2026
613.00
Save 91.35 (15%)
Each
Chemical Identifiers
CAS87583-89-9
IUPAC NameN1,N2-dimethylcyclohexane-1,2-diamine
Molecular FormulaC8H18N2
InChI KeyJRHPOFJADXHYBR-UHFFFAOYNA-N
SMILESCNC1CCCCC1NC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
It is employed in the synthesis of 6-substituted 1-oxoindanoyl isoleucine conjugates and modeling studies with the coi1-jaz co-receptor complex of lima bean.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of 6-substituted 1-oxoindanoyl isoleucine conjugates and modeling studies with the coi1-jaz co-receptor complex of lima bean.

Solubility
Slightly soluble in water.

Notes
Store away from acids, air, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Store under inert gas. Keep it refrigerated.
RUO – Research Use Only

General References:

  1. Yoko Nakamura, et al. Synthesis of 6-Substituted 1-oxoindanoyl Isoleucine Conjugates and Modeling Studies with the COI1-JAZ Co-Receptor Complex of Lima Bean.J. Chem. Ecol.,2014,407(7), 687-699.
  2. Rebecca M. Steele, et al. Enantioselective cyanosilylation of aldehydes catalyzed by a diastereomeric mixture of atropisomeric thioureas.Tetrahedron: Asymmetry.,2006,17(6), 999-1006.