2-[4-(Trifluoromethyl)phenyl]ethylamine, 98%
2-[4-(Trifluoromethyl)phenyl]ethylamine, 98%
2-[4-(Trifluoromethyl)phenyl]ethylamine, 98%
2-[4-(Trifluoromethyl)phenyl]ethylamine, 98%
Thermo Scientific Chemicals

2-[4-(Trifluoromethyl)phenyl]ethylamine, 98%

CAS: 775-00-8 | C9H10F3N | 189.181 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64415.06
also known as H64415-06
Price (USD)
234.65
Online Exclusive
261.00
Save 26.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
234.65
Online Exclusive
261.00
Save 26.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS775-00-8
IUPAC Name2-[4-(trifluoromethyl)phenyl]ethan-1-aminium
Molecular FormulaC9H11F3N
InChI KeyHMFOBPNVAAAACP-UHFFFAOYSA-O
SMILES[NH3+]CCC1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
It is applied as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only