5-Cyanothiophene-2-carboxaldehyde is found in caper and may have pest control activity against meloidogyne incognita.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Cyanothiophene-2-carboxaldehyde is found in caper and may have pest control activity against meloidogyne incognita.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
Pierluigi Caboni; Giorgia Sarais; Nadhem Aissani; Graziella Tocco; Nicola Sasanelli; Barbara Liori; Annarosa Carta; and Alberto Angioni. Nematicidal Activity of 2-Thiophenecarboxaldehyde and Methylisothiocyanate from Caper (Capparis spinosa) against Meloidogyne incognita. J. Agric. Food Chem. 2012 60(30), 7345-7351.