3,5-Bis(methoxycarbonyl)benzeneboronic acid, 97%
3,5-Bis(methoxycarbonyl)benzeneboronic acid, 97%
3,5-Bis(methoxycarbonyl)benzeneboronic acid, 97%
Thermo Scientific Chemicals

3,5-Bis(methoxycarbonyl)benzeneboronic acid, 97%

CAS: 177735-55-6 | C10H11BO6 | 238.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64472.06
also known as H64472-06
Price (USD)
816.65
Online Exclusive
907.00
Save 90.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
816.65
Online Exclusive
907.00
Save 90.35 (10%)
Each
Chemical Identifiers
CAS177735-55-6
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
It is employed in the for the synthesis of 3,4-disubstituted coumarins via Pd-Catalyzed site-selective cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the for the synthesis of 3,4-disubstituted coumarins via Pd-Catalyzed site-selective cross-coupling reactions.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only
Liang Zhang.; Tianhao Meng.; Renhua Fan.; Jie Wu. General and Efficient Route for the Synthesis of 3,4-Disubstituted Coumarins via Pd-Catalyzed Site-Selective Cross-Coupling Reactions. J. Org. Chem. 2007, 72 (19),7279-7286 .