1,5-Naphthyridine, 97+%
1,5-Naphthyridine, 97+%
1,5-Naphthyridine, 97+%
Thermo Scientific Chemicals

1,5-Naphthyridine, 97+%

CAS: 254-79-5 | C8H6N2 | 130.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H64484.06
also known as H64484-06
Price (USD)
510.65
Online Exclusive
567.00
Save 56.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
510.65
Online Exclusive
567.00
Save 56.35 (10%)
Each
Chemical Identifiers
CAS254-79-5
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only