2,6-Dimethylbenzaldehyde, 97%
2,6-Dimethylbenzaldehyde, 97%
2,6-Dimethylbenzaldehyde, 97%
2,6-Dimethylbenzaldehyde, 97%
Thermo Scientific Chemicals

2,6-Dimethylbenzaldehyde, 97%

CAS: 1123-56-4 | C9H10O | 134.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H64503.14
also known as H64503-14
Price (USD)
144.65
Special offer
Online exclusive
Ends: 30-Jun-2026
170.00
Save 25.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
144.65
Special offer
Online exclusive
Ends: 30-Jun-2026
170.00
Save 25.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS1123-56-4
IUPAC Name2,6-dimethylbenzaldehyde
Molecular FormulaC9H10O
InChI KeyQOJQBWSZHCKOLL-UHFFFAOYSA-N
SMILESCC1=CC=CC(C)=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
FormLow melting solid
Appearance (Color)White to yellow
An aldehyde containing building block used for proteomics research

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An aldehyde containing building block used for proteomics research

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Hing L. Sham; David A. Betebenner; Xiaoqi Chen; Ayda Saldivar; Sudthida Vasavanonda; Dale J. Kempf; Jacob J. Plattner; Daniel W. Norbeck. Synthesis and structure-activity relationships of a novel series of HIV-1 protease inhibitors encompassing ABT-378 (Lopinavir). Bioorganic & Medicinal Chemistry Letters. 1972 12(8), 1185-1187.
  2. C. L. Bell; S. S. Danyluk; T. Schaefer.Signs of spin-spin coupling constants between aldehydic and ring protons in 2,6-dinitrobenzaldehyde. Evidence for a hyperconjugative contribution to JpH,CHO. Canadian Journal of Chemistry. 1969 47(19), 3529-3533.