6-Hydroxy-1,2,3,4-tetrahydroisoquinoline, 97%
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline, 97%
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline, 97%
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline, 97%
Thermo Scientific Chemicals

6-Hydroxy-1,2,3,4-tetrahydroisoquinoline, 97%

CAS: 14446-24-3 | C9H11NO | 149.193 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H64512.MD
also known as H64512-MD
Price (USD)
129.65
Online Exclusive
144.00
Save 14.35 (10%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
129.65
Online Exclusive
144.00
Save 14.35 (10%)
Each
Add to cart
Specifications
Beilstein127578
CAS14446-24-3
Chemical Name or Material6-Hydroxy-1,2,3,4-tetrahydroisoquinoline
Formula Weight149.19
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only