2-Bromodibenzothiophene, 98%
2-Bromodibenzothiophene, 98%
2-Bromodibenzothiophene, 98%
Thermo Scientific Chemicals

2-Bromodibenzothiophene, 98%

CAS: 22439-61-8 | C12H7BrS | 263.152 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H64645.14
also known as H64645-14
Price (USD)
426.65
Special offer
Online exclusive
Ends: 30-Jun-2026
502.00
Save 75.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
426.65
Special offer
Online exclusive
Ends: 30-Jun-2026
502.00
Save 75.35 (15%)
Each
Chemical Identifiers
CAS22439-61-8
IUPAC Name4-bromo-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular FormulaC12H7BrS
InChI KeyIJICRIUYZZESMW-UHFFFAOYSA-N
SMILESBrC1=CC2=C(SC3=C2C=CC=C3)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
Used as intermediate for organic light-emitting diode(OLED) materials and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediate for organic light-emitting diode(OLED) materials and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Henry Gilman; Donald L. Esmay. The Cleavage of Heterocycles with Raney Nickel and with Lithium. J. Am. Chem. Soc. 1953 75(12), 2947-2949.
  2. Osamu Shimomura; Toshihiko Sato; Ikuyoshi Tomita; Masato Suzuki; and Takeshi Endo. Synthesis and radical polymerization of 2-vinyldibenzothiophene. Journal of Polymer Science Part A: Polymer Chemistry. 1997 35(14), 2813-2819.