tert-Butyl hydrogen succinate, 97%
tert-Butyl hydrogen succinate, 97%
tert-Butyl hydrogen succinate, 97%
tert-Butyl hydrogen succinate, 97%
Thermo Scientific Chemicals

tert-Butyl hydrogen succinate, 97%

CAS: 15026-17-2 | C8H14O4 | 174.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64706.03
also known as H64706-03
Price (USD)
38.50
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
38.50
Each
Add to cart
Chemical Identifiers
CAS15026-17-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow-brown
FormCrystalline powder
Assay (GC)>96.0%
It is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Alain Dhainaut; Gilbert Regnier; Ghanem Atassi; Alain Pierre; Stephane Leonce; Laurence Kraus-Berthier; Jean Francois Prost. New triazine derivatives as potent modulators of multidrug resistance. J. Med. Chem.1992 35(13), 2481-2496.
  2. Rebekah Beevers; Maria G Carr; Philip S Jones; Steven Jordan; Paul B Kay; Robert C Lazell; Tony M Raynham.Solution and solid-Phase synthesis of potent inhibitors of hepatitis C Virus NS3 proteinase. Bioorganic & Medicinal Chemistry Letters.2002 12(4), 641-643.