2-(Chloromethyl)benzothiazole, 95%
2-(Chloromethyl)benzothiazole, 95%
2-(Chloromethyl)benzothiazole, 95%
2-(Chloromethyl)benzothiazole, 95%
Thermo Scientific Chemicals

2-(Chloromethyl)benzothiazole, 95%

CAS: 37859-43-1 | C8H6ClNS | 183.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64781.03
also known as H64781-03
Price (USD)
108.65
Online Exclusive
120.00
Save 11.35 (9%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
108.65
Online Exclusive
120.00
Save 11.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS37859-43-1
IUPAC Name2-(chloromethyl)-1,3-benzothiazole
Molecular FormulaC8H6ClNS
InChI KeySERUZNHRWBXDOX-UHFFFAOYSA-N
SMILESClCC1=NC2=CC=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only