2-Bromo-4-fluoropyridine, 97%
2-Bromo-4-fluoropyridine, 97%
2-Bromo-4-fluoropyridine, 97%
Thermo Scientific Chemicals

2-Bromo-4-fluoropyridine, 97%

CAS: 357927-50-5 | C5H3BrFN | 175.988 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64785.06
also known as H64785-06
Price (USD)
188.65
Special offer
Online exclusive
Ends: 30-Jun-2026
222.00
Save 33.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
188.65
Special offer
Online exclusive
Ends: 30-Jun-2026
222.00
Save 33.35 (15%)
Each
Chemical Identifiers
CAS357927-50-5
IUPAC Name2-bromo-4-fluoropyridine
Molecular FormulaC5H3BrFN
InChI KeyAIEATTRZKVGMBO-UHFFFAOYSA-N
SMILESFC1=CC(Br)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only