Pyrazine-2,5-dicarboxylic acid, 95%
Pyrazine-2,5-dicarboxylic acid, 95%
Pyrazine-2,5-dicarboxylic acid, 95%
Thermo Scientific Chemicals

Pyrazine-2,5-dicarboxylic acid, 95%

CAS: 122-05-4 | C6H4N2O4 | 168.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64795.06
also known as H64795-06
Price (USD)
243.65
Special offer
Online exclusive
Ends: 30-Jun-2026
286.00
Save 42.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
243.65
Special offer
Online exclusive
Ends: 30-Jun-2026
286.00
Save 42.35 (15%)
Each
Chemical Identifiers
CAS122-05-4
IUPAC Namepyrazine-2,5-dicarboxylic acid
Molecular FormulaC6H4N2O4
InChI KeyGMIOYJQLNFNGPR-UHFFFAOYSA-N
SMILESOC(=O)C1=CN=C(C=N1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
It is an pyrazine used for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an pyrazine used for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. J.S. Brown; M.J.S. Dewa. Thermal and electrical properties of transition metal complexes and coordination polymers of pyrazine-2,5-dicarboxylic acid. Inorganica Chimica Acta.1979 34221-224.
  2. W. J. Schut; H. I. X. Mager; and W. Berends. Investigations on pyrazine derivatives. VII: On the conversion of pyrazine 2,5-dicarboxylic acid into some derivatives of 2-aminopyrazine 5-carboxylic acid. Recueil des Travaux Chimiques des Pays-Bas.1961 80(4), 391-398.