2-Amino-4-nitrobenzoic acid, 97+%
2-Amino-4-nitrobenzoic acid, 97+%
2-Amino-4-nitrobenzoic acid, 97+%
Thermo Scientific Chemicals

2-Amino-4-nitrobenzoic acid, 97+%

CAS: 619-17-0 | C7H6N2O4 | 182.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H64831.22
also known as H64831-22
Price (USD)
104.65
Special offer
Online exclusive
Ends: 30-Jun-2026
123.00
Save 18.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
104.65
Special offer
Online exclusive
Ends: 30-Jun-2026
123.00
Save 18.35 (15%)
Each
Chemical Identifiers
CAS619-17-0
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only