4-(Trifluoromethyl)piperidine, 97%
4-(Trifluoromethyl)piperidine, 97%
4-(Trifluoromethyl)piperidine, 97%
Thermo Scientific Chemicals

4-(Trifluoromethyl)piperidine, 97%

CAS: 657-36-3 | C6H10F3N | 153.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64840.03
also known as H64840-03
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS657-36-3
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only