3-Aminopyridazine, 97%
3-Aminopyridazine, 97%
3-Aminopyridazine, 97%
Thermo Scientific Chemicals

3-Aminopyridazine, 97%

CAS: 5469-70-5 | C4H5N3 | 95.105 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H64859.03
also known as H64859-03
Price (USD)
38.60
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
38.60
Each
Chemical Identifiers
CAS5469-70-5
IUPAC Namepyridazin-3-amine
Molecular FormulaC4H5N3
InChI KeyLETVJWLLIMJADE-UHFFFAOYSA-N
SMILESNC1=CC=CN=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormSolid
Assay (HPLC)>96.0%
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only
Camille Georges Wermuth.; Gilbert Schlewer.; Jean Jacques Bourguignon.; Georges Maghioros.; Marie Jeanne Bouchet.; Claudine Moire.; Jean Paul Kan.; Paul Worms.; Kathleen Biziere. 3-Aminopyridazine derivatives with atypical antidepressant, serotonergic and dopaminergic activities.J. Med. Chem. 1989, 32 (3),528-537 .